Learn More
ZeptoMetrix Corporation HBV Longitudinal Panels Available on GSA/VA Contract for Federal Government customers only.
collected from donors that are negative prior to the vaccination and positive after the vaccination
Manufacturer:  ZeptoMetrix CorporationSupplier Diversity Partner 6508
 View more versions of this product
Catalog No. 22-156-914
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
17 year old caucasian male
HBV longitudinal reference panel over 31 weeks
15 x 1mL
-20°C