Learn More
ZeptoMetrix Corporation HBV Seroconversion Panels - 20 to 40 Member Panel
Donor Demographics: Donor source plasma was collected from a U.S. licensed blood collection facility. Donor was un-treated and asymptomatic throughout the collection period.
Manufacturer: ZeptoMetrix Corporation HBV6272
Specifications
-20°C | |
HBV seroconversion reference panel | |
26 x 1mL | |
FDA |