Learn More
ZeptoMetrix Herpes Simplex Virus (HSV) Type 1 Culture Fluid
Infectious virus preparation designed to meet research and product development needs
Supplier: ZeptoMetrix 0810200CF
Alert:
Contains live and infectious organisms.
Description
Human Herpes viruses are enveloped icosahedral viruses 150 to 200nm in diameter and contain a nucleocapsid and double-stranded linear DNA.
- Consist of virus, cells and media taken directly from tissue culture flask
- Propagated in Vero cell line
- Biosafety Level 2 organism
- Available in 1.0mL aliquots
- Each lot assayed for its Tissue Culture Infective Dose (TCID50)
- Titer:>105 U/mL
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Nucleic acid/molecular testing, limit of detection (LOD) studies, cross-reactivity studies and other viral-based assays
Specifications
2 | |
>105U/mL | |
Solution | |
Herpes Simplex Virus Infectious Culture Fluid | |
1 mL |
ISO, 13485 | |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Safety and Handling
ShelfLife : 1 Year
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.