Learn More
ZeptoMetrix Herpes Simplex Virus (HSV) Type 1 Culture Fluid
Infectious virus preparation designed to meet research and product development needs
$380.00 - $435.00
Specifications
Biosafety Level | 2 |
---|---|
Certifications/Compliance | ISO, 13485 |
Concentration | >105U/mL |
Content And Storage | -65°C |
Form | Solution |
Alert:
Contains live and infectious organisms.
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
22-157-549
|
ZeptoMetrix
0810005CFN |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $389.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-548
|
ZeptoMetrix
0810005CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $380.00
|
|
|||||
22-157-719
|
ZeptoMetrix
0810183CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-720
|
ZeptoMetrix
0810184CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-721
|
ZeptoMetrix
0810185CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-723
|
ZeptoMetrix
0810187CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-722
|
ZeptoMetrix
0810186CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
|
|||||
22-157-724
|
ZeptoMetrix
0810188CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-725
|
ZeptoMetrix
0810189CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-727
|
ZeptoMetrix
0810191CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-726
|
ZeptoMetrix
0810190CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-728
|
ZeptoMetrix
0810192CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-730
|
ZeptoMetrix
0810194CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-731
|
ZeptoMetrix
0810195CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $2,307.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-729
|
ZeptoMetrix
0810193CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-733
|
ZeptoMetrix
0810197CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-732
|
ZeptoMetrix
0810196CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-734
|
ZeptoMetrix
0810198CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-737
|
ZeptoMetrix
0810201CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-735
|
ZeptoMetrix
0810199CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-736
|
ZeptoMetrix
0810200CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
|
|||||
22-157-738
|
ZeptoMetrix
0810202CF |
Herpes Simplex Virus Infectious Culture Fluid |
Each for $435.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Human Herpes viruses are enveloped icosahedral viruses 150 to 200nm in diameter and contain a nucleocapsid and double-stranded linear DNA.
- Consist of virus, cells and media taken directly from tissue culture flask
- Propagated in Vero cell line
- Biosafety Level 2 organism
- Available in 1.0mL aliquots
- Each lot assayed for its Tissue Culture Infective Dose (TCID50)
- Titer:>105 U/mL
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Nucleic acid/molecular testing, limit of detection (LOD) studies, cross-reactivity studies and other viral-based assays
Specifications
2 | |
>105U/mL | |
Solution | |
Herpes Simplex Virus Infectious Culture Fluid | |
1 mL |
ISO, 13485 | |
-65°C | |
Vial | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Safety and Handling
ShelfLife : 1 Year
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.