Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Corporation Human Immunodeficiency Virus Type 1 (HIV-1) Culture Fluid
Infectious virus preparation designed to meet research and product development needs
Manufacturer: ZeptoMetrix Corporation 0810028CF
Alert:
Contains live and infectious organisms.
Description
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
2 | |
Human immunodeficiency virus type 1 infectious culture fluid | |
BaL | |
1mL | |
Vial |
-65°C | |
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays | |
1 Year | |
Solution | |
ISO, 13485 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title