Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Human Immunodeficiency Virus Type 1 (HIV-1) Culture Fluid

Infectious virus preparation designed to meet research and product development needs

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Media Type Vial
View More Specs

Alert:

Contains live and infectious organisms.


Products 3
Catalog Number Mfr. No. Strain Designations Price Quantity & Availability  
Catalog Number Mfr. No. Strain Designations Price Quantity & Availability  
22-157-485
View Documents
ZeptoMetrix Corporation
0801032CF
IIIB
Each for $317.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-578
View Documents
ZeptoMetrix Corporation
0810027CF
Mn
Each for $317.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-580
View Documents
ZeptoMetrix Corporation
0810028CF
BaL
Each for $317.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Specifications

2
Solution
Vial
1 Year
ISO, 13485
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1mL
-65°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit