Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Staphylococcus aureus MSSA; δmecA, Genomic DNA

For use as an amplification and/or detection control for nucleic acid testing of S. aureus

Specifications

Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Storage Requirements -20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-525
View Documents
ZeptoMetrix Corporation
0801675DNA10UG
10μg
Each for $436.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Can also be used to determine a limit of detection (LOD), in diagnostic assay development, cross-reactivity studies or genomic sequencing
  • Each aliquot contains DNA extracted from pure culture
  • Supplied in TE Buffer

Amplification/detection control for nucleic acid testing of Staphylococcus aureus

Specifications

Specifications

ISO, 13485
Solution
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
-20°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit