ZeptoMetrix Corporation Staphylococcus aureus MSSA; δmecA, Genomic DNA Available on GSA/VA Contract for Federal Government customers only.

For use as an amplification and/or detection control for nucleic acid testing of S. aureus

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner 0801675DNA100UG

 View more versions of this product

Catalog No. 22-157-524

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
    Format Solution
    Product Type Staphylococcus aureus MSSA deltamecA clinical isolate genomic DNA bacteria control
    Quantity 100μg
    Storage Requirements -20°C
    Certifications/Compliance ISO, 13485

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit