Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Tremella fuciformis-S cerevisiae Recombinant Extraction Control

Each frozen aliquot contains 1mL of pure, titered culture of a recombinant strain of Shiga Toxin 2-S.cerevisiae

Specifications

Biosafety Level 1
Certifications/Compliance ISO, 13485
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Storage Requirements -65°C
Format Solution
View More Specs

Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-157-386
View Documents
ZeptoMetrix Corporation
0801802
Tremella fuciformis-Saccharomyces cerevisiae titered
Each for $432.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Biosafety level 1 microorganism
  • Recombinant, Titered Form:
  • Can be used to determine a limit of detection (LOD), in diagnostic assay development or cross-reactivity studies
  • Recombinant, Fixed and Titered Form:
  • Can be used as an internal control in nucleic acid-based amplification assays

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Specifications

Specifications

1
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Solution
ISO, 13485
-65°C
1mL
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit