Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Influenza A H1N1 2009 (Swine Flu) Culture Fluid

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Media Type Vial
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-157-669
View Documents
ZeptoMetrix Corporation
0810109CFJ
Canada isolate
Each for $316.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-670
View Documents
ZeptoMetrix Corporation
0810109CFN
NY isolate
Each for $316.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

IMPORTANT: Contains live material, so cust must submit an executed ZeptoMetrix MTA before vendor can ship. Also, a USDA APHIS VS 16-6 or 16-6A authorized permit must be obtained and a valid copy of the permit received by ZeptoMetrix prior to shipment of this product. requests.

Specifications

Specifications

2
Solution
Vial
1 Year
ISO, 13485
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1mL
-65°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit