Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Mumps Virus Culture Fluid

Infectious virus preparation designed to meet research and product development needs

Specifications

Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Media Type Vial
Quantity 1mL
View More Specs

Alert:

Contains live and infectious organisms.


Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
22-157-633
View Documents
ZeptoMetrix Corporation
0810079CF
Isolate 1
Each for $307.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-718
View Documents
ZeptoMetrix Corporation
0810176CF
Isolate 2
Each for $307.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Nucleic acid /molecular testing, limit of detection (LOD) studies, cross-reactivity studies, other viral-based assays

Specifications

Specifications

ISO, 13485
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1mL
-65°C
Solution
Vial
1 Year
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit