Learn More
ZeptoMetrix Corporation NATtrol™ Herpes Simplex Virus Type 1, MacIntyre Strain
Designed to evaluate the performance of nucleic acid tests for determination of the presence of HSV1 DNA
Manufacturer:  ZeptoMetrix Corporation NATHSV10001
 View more versions of this product
Catalog No. 22-156-897
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 1.0mL vials of target concentrations (100, 500, 1000, 10000 and 50000 copies/mL)
NATHSV1 contains intact virus and should be run in a manner identical to that used for clinical specimens
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
2° to 8°C
100 copies/mL
1mL
Vial
NATtrol™ Herpes Simplex virus type 1 MacIntyre strain virus control
Determination of presence of HSV1 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Solution
ISO, 13485