Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Herpes Simplex Virus Type 2, MS Strain
Designed to evaluate the performance of nucleic acid tests for determination of the presence of HSV2 DNA.
Supplier: ZeptoMetrix NATHSV20003
Description
- Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Contains 1.0mL vials of target concentrations (100, 500, 1000, 10000 and 50000 copies/mL)
NATHSV2 contains intact virus and should be run in a manner identical to that used for clinical specimens.
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
ISO, 13485 | |
2°C to 8°C | |
Vial | |
Determination of presence of HSV2 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
1000 copies/mL | |
Solution | |
NATtrol Virus Control | |
1 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction