Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation NATtrol™ Herpes Simplex Virus Type 2, MS Strain Available on GSA/VA Contract for Federal Government customers only.

Designed to evaluate the performance of nucleic acid tests for determination of the presence of HSV2 DNA.

$46.00 - $482.00

Specifications

Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Determination of presence of HSV2 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Media Type Vial
Quantity 1mL
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
22-156-746 Available on GSA/VA Contract for Federal Government customers only.
View Documents
ZeptoMetrix Corporation
NATHSV20003
1000 copies/mL
Each for $99.00
Add to cart
 
22-156-745
View Documents
ZeptoMetrix Corporation
NATHSV20002
500 copies/mL
Each for $46.00
Add to cart
 
22-156-747 Available on GSA/VA Contract for Federal Government customers only.
View Documents
ZeptoMetrix Corporation
NATHSV20005
50000 copies/mL
Each for $482.00
Add to cart
 
22-156-899
View Documents
ZeptoMetrix Corporation
NATHSV20001
100 copies/mL
Each for $36.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-156-900 Available on GSA/VA Contract for Federal Government customers only.
View Documents
ZeptoMetrix Corporation
NATHSV20004
10000 copies/mL
Each for $285.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 1.0mL vials of target concentrations (100, 500, 1000, 10000 and 50000 copies/mL)

NATHSV2 contains intact virus and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

ISO, 13485
Determination of presence of HSV2 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
1mL
Solution
Vial
2° to 8°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit