Learn More
ZeptoMetrix Corporation NATtrol™ Herpes Simplex Virus Type 2, MS Strain Available on GSA/VA Contract for Federal Government customers only.
Designed to evaluate the performance of nucleic acid tests for determination of the presence of HSV2 DNA.
Manufacturer:  ZeptoMetrix CorporationSupplier Diversity Partner NATHSV20005
 View more versions of this product
Catalog No. 22-156-747
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ISO, 13485
Determination of presence of HSV2 DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Vial
1mL
50000 copies/mL
Solution
NATtrol™ Herpes Simplex virus type 2 MS strain virus control
2° to 8°C