Learn More
ZeptoMetrix Corporation NATtrol™ Human Parvovirus, B19 Strain Available on GSA/VA Contract for Federal Government customers only.
Designed to evaluate the performance of nucleic acid tests for determination of the presence of Parvovirus DNA
Manufacturer:  ZeptoMetrix CorporationSupplier Diversity Partner NATPARVO0004
 View more versions of this product
Catalog No. 22-156-902
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
ISO, 13485
Determination of presence of Parvovirus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Formulated with purified, intact virus particles
NATtrol™ Human Parvovirus B19 strain control
2° to 8°C
10000000IU/mL
Solution
Vial
1mL
Safety and Handling
ShelfLife : 1 Year