ZeptoMetrix Corporation NATtrol™ Human Parvovirus, B19 Strain Available on GSA/VA Contract for Federal Government customers only.

Designed to evaluate the performance of nucleic acid tests for determination of the presence of Parvovirus DNA

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner NATPARVO0003

 View more versions of this product

Catalog No. 22-156-754

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Concentration 1000000IU/mL
    For Use With (Application) Determination of presence of Parvovirus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
    Format Solution
    Formulation Formulated with purified, intact virus particles
    Media Type Vial
    Product Type NATtrol™ Human Parvovirus B19 strain control
    Quantity 1mL
    Shelf Life 1 Year
    Storage Requirements 2° to 8°C
    Certifications/Compliance ISO, 13485

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit