Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation NATtrol™ Escherichia coli External Run Control

Supplied in a purified serum protein matrix that mimics the composition of a true clinical specimen

Specifications

Certifications/Compliance ISO, 13485
Format Solution
Media Type Vial
Formulation Formulated with purified, intact bacterial cells
Quantity 6 x 1mL
View More Specs

Products 2
Catalog Number Mfr. No. Concentration For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Concentration For Use With (Application) Price Quantity & Availability  
22-157-781
View Documents
ZeptoMetrix Corporation
NATECO(933)ERCL
5000cfu/mL Extraction and amplification of Escherichia coli, Low, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Each for $335.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-782
View Documents
ZeptoMetrix Corporation
NATECO(933)ERCM
50000cfu/mL Extraction and amplification of Escherichia coli, medium, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Each for $422.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Formulated with intact bacteria that are chemically modified to render them non-infectious and refrigerator stable
  • Calibrated against an in-house DNA standard
  • Contains 6 x 1mL vials
  • Available concentrations: 5000 or 50,000 cfu/mL

Extraction and amplification of E. coli

Specifications

Specifications

ISO, 13485
Vial
6 x 1mL
Solution
Formulated with purified, intact bacterial cells
2° to 8°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit