Learn More
ZeptoMetrix NATtrol™ Influenza A Virus
Qualitative molecular control
$1678.00
Specifications
Content And Storage 2°C to 8°C
Quantity 1 mL
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-156-736
View Documents
ZeptoMetrix
NATFLUAH3ST
Influenza A H3 Virus
Each for $1,678.00
Add to Cart
22-156-891
View Documents
ZeptoMetrix
NATFLUAH1ST
Influenza A H1 Virus
Each for $1,678.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Specifications
2°C to 8°C
1 mL