Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Influenza B External Run Controls Available on GSA/VA Contract for Federal Government customers only.

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

$421.00

Specifications

Detectable Analytes Determination of presence of Influenza B RNA
Product Type Influenza B External Run Control
Quantity 6 x 1mL
Storage Requirements 2°C to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Price Quantity & Availability  
22-156-737
View Documents
ZeptoMetrix
NATFLUBERCM
25 to 28 Ct
Each for $421.00
Add to cart
 
22-156-892 Available on GSA/VA Contract for Federal Government customers only.
View Documents
ZeptoMetrix
NATFLUBERCL
28 to 31 Ct
Each for $346.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Each control pack contains 6 x 1.0mL vials of Influenza B NATtrol™ at concentrations of 25 to 31 Ct Range
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate performance of nucleic acid tests for determination of presence of Influenza B RNA

Contain intact virus and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Specifications

Determination of presence of Influenza B RNA
Influenza B External Run Control
6 x 1mL
2°C to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit