Learn More
ZeptoMetrix Corporation NATtrol™ Respiratory Validation Panel Available on GSA/VA Contract for Federal Government customers only.
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
Manufacturer:  ZeptoMetrix CorporationSupplier Diversity Partner NATRVP3
 View more versions of this product
Catalog No. 22-156-757
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
ISO 9001:2008, ISO 13485:2003
Solution
NATtrol™ respiratory validation panel
2° to 8°C
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Vial
17 x 0.2mL