ZeptoMetrix Corporation NATtrol™ Respiratory Validation Panel Available on GSA/VA Contract for Federal Government customers only.

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner NATRVP1

 View more versions of this product

Catalog No. 22-156-755

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
    Format Solution
    Media Type Vial
    Product Type NATtrol™ respiratory validation panel
    Quantity 13 x 0.2mL
    Storage Requirements 2° to 8°C
    Certifications/Compliance ISO 9001:2008, ISO 13485:2003

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit