Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Respiratory Validation Panel

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

Specifications

Certifications/Compliance ISO 9001:2008, ISO 13485:2003
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Media Type Vial
Storage Requirements 2° to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-156-757
View Documents
ZeptoMetrix
NATRVP3
17 x 0.2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. View substitute products »
22-156-755
View Documents
ZeptoMetrix
NATRVP1
13 x 0.2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Controls supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate the performance of nucleic acid tests for determination of the presence of viral nucleic acids

Panel Members:

  • Negative
  • Influenza A H1
  • Influenza A H3
  • Influenza B
  • Respiratory Syncytial Virus A
  • Respiratory Syncytial Virus B
  • Parainfluenza virus Type 1
  • Parainfluenza virus Type 2
  • Parainfluenza virus Type 3
  • Adenovirus Type 3
  • Enterovirus
  • Human Metapneumovirus
  • Coronavirus 229E
  • Coronavirus OC43
  • Coronavirus SARS
  • Influenza A H1N1 (2009)

NATRVP-1

  • Contains 13 x 0.2mL vials each containing viral NATtrol™ targets of panel members
NATRVP-2
  • Contains 16 x 0.2mL vials each containing viral NATtrol™ targets of panel members
NATRVP-3
  • Contains 17 x 0.2mL vials each containing viral NATtrol™ targets of panel members

IMPORTANT: Contains Metapneumovirus. Before vendor can ship order, customer must submit a Certificate of Compliance (COC) regarding use restrictions.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Specifications

ISO 9001:2008, ISO 13485:2003
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
2° to 8°C
Solution
Vial
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit