Learn More
ZeptoMetrix NATtrol™ RP Multimarker Control Pack
Formulated with purified, intact organisms that have been chemically modified to render them non-infectious and refrigerator stable
Supplier: ZeptoMetrix MDZ001
Description
- Contains 3 x 0.75mL vials of RPMultimarker 1 and 3 x 0.75mL vials RP Multimarker 2
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
2°C to 8°C | |
RP Multimarker Control | |
6 x 0.75 mL |
Solution | |
Multiplex, respiratory molecular assays |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
This product is an IVD/CE marked USA FDA Class 1 exempt, unassayed external run control intended for in vitro diagnostic use and are for professional use only. NATtrol™ controls are not intended for use as a substitute for the internal controls provided by in vitro diagnostic kit manufacturers. Quality control materials should be used in accordance with local, state, federal and accreditation requirements.