ZeptoMetrix Corporation NATtrol™ Varicella-Zoster Virus, Strain: Ellen Available on GSA/VA Contract for Federal Government customers only.

Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner NATVZV0003

 View more versions of this product

Catalog No. 22-156-763

  Check Availability
Add to Cart

    Specifications

    Quantity 1mL
    Concentration 1000 copies/mL
    For Use With (Application) Determination of presence of VZV RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
    Format Solution
    Formulation Formulated with purified, intact virus particles
    Media Type Vial
    Product Type NATtrol™ Varicella Zoster virus Ellen strain control
    Shelf Life 2 Years
    Storage Requirements 2° to 8°C
    Certifications/Compliance ISO, 13485

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit