Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ West Nile Virus, Strain: NY 2001-6263
Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
Supplier: ZeptoMetrix NATWNV0001
Description
- Each vial contains 1.0mL of WNV NATtrol at various concentrations
- Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
- Can be used to evaluate the performance of nucleic acid tests for determination of the presence of WNV RNA
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
ISO, 13485 | |
2°C to 8°C | |
Vial | |
Formulated with purified, intact virus particles | |
1 mL |
100 copies/mL | |
Solution | |
NATtrol Virus Strain Control | |
Determination of presence of WNV RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
Safety and Handling
ShelfLife : 2 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction