Learn More
ZeptoMetrix Purified Viral Lysate, Herpes Simplex Virus Type-2 (HSV-2)
Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies
Supplier: ZeptoMetrix 0810006
Description
- Concentration ranges from 0.5 to 3.0mg/mL
- Available in 1mg vials
Immunoassays, Western Blot, Dot Blot and other protein-based assays
Specifications
2 | |
0.5 to 3mg/mL | |
Viral Antibodies | |
Vial | |
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay | |
1 mg |
ISO, 13485 | |
-65°C | |
Solution | |
Purified Viral Lysate | |
Sucrose density gradient ultracentrifugation |
Safety and Handling
ShelfLife : 1 Year