Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation Purified Viral Lysate, Human Immunodeficiency Virus Type 1 (HIV-1)

Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Detects Viral Antibodies
Concentration 0.5 to 3mg/mL
Format Solution
View More Specs

Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-157-412
View Documents
ZeptoMetrix Corporation
0810028
Human immunodeficiency virus type 1 BaL strain purified viral lysate
Each for $1,486.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-411
View Documents
ZeptoMetrix Corporation
0810027
Human immunodeficiency virus type 1 Mn strain purified viral lysate
Each for $1,486.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Available in 1mg vials

Specifications

Specifications

2
Viral Antibodies
Solution
Vial
1mg
-65°C
ISO, 13485
0.5 to 3mg/mL
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay
Sucrose density gradient ultracentrifugation
1 Year
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit