Learn More
ZeptoMetrix Corporation Purified Viral Lysate, West Nile Virus (WNV)
Can be utilized as antigen, source for purification of viral proteins or for detection of viral antibodies
Manufacturer: ZeptoMetrix Corporation 0810082
Description
- Available in 1mg vials
Specifications
2 | |
Sucrose density gradient ultracentrifugation | |
West Nile virus 2 (1986) strain purified viral lysate | |
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay | |
1mg | |
Vial |
Viral Antibodies | |
-65°C | |
0.5 to 3mg/mL | |
1 Year | |
Solution | |
ISO, 13485 |