Learn More
ZeptoMetrix Corporation Recombinant Human Growth Factors
Mediates the process of clonal expansion and maintenance of T-cell proliferation
Manufacturer: ZeptoMetrix Corporation 0802001
Description
CELLKINES rhIL-2 originates from the IL-2 gene found in human Jurkat cell line which was cloned into E. coli. This product differs from native IL-2 in two ways. First, it is not glycosylated, and second, the protein differs at two amino acid positions (the first three N-terminal amino acids are absent, and CYS at position 125 is substituted by SER). CELLKINES rhIL-2 with a molecular weight of 15.4 kDa is formulated in 2% fetal bovine serum (heat inactivated) as a stabilizer and sterile filtered.
- High specific activity and purity
Specifications
100μg | |
-20°C | |
Recombinant Human Interleukin-2 | |
Research use only |
1 Year | |
Human Jurkat Cell Line | |
E. coli |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.