Learn More
ZeptoMetrix RETROtek HIV-1 p24 Antigen 2.0 ELISA
Detects Human Immunodeficiency Virus Type 1 (HIV-1) p24 antigen in cell culture media
Supplier: ZeptoMetrix 0801008
Alert:
Contains sodium azide as a preservative.
Includes: 5 plates of HIV-1 p24 Antibody Coated Microplate for 96 determinations, 12 x 8 well strips, 5 x 12mL of HIV-1 p24 detector antibody, 5 x 0.5mL of HIV-1 p24 antigen standard, 5 x 5mL of lysing buffer, 5 x 100mL of assay diluent, 5 x 125mL of 10X plate wash buffer, 5 x 12mL of substrate, 5 x 12mL of stop solution, 5 x 10 plate sealers, 5 resealable plastic bag
Description
- This assay detects p24 from HIV-1 clades A-F, since the amino acid sequence of p24 is well conserved
- There is no cross reactivity to Human Immunodeficiency Virus Type-2 (HIV-2), Simian Immunodeficiency Virus (SIV) or Human T-Cell Leukemia Virus Types I and II (HTLV I and II) p24 or any gag gene product
- The assay can be used to monitor the purification and biochemical behavior of HIV-1 or determine the titer of HIV-1-based lentiviral samples
Specifications
RETROtek HIV-1 p24 Antigen 2.0 ELISA | |
Used to detect human immunodeficiency virus type 1 (HIV-1) p24 antigen in cell culture media, It can be used to monito r the purification and biochemical behavior of HIV-1 or determine the titer of HIV-1-based lentiviral samples, since the amino acid sequence of p24 is well conserved among the various HIV-1 isolates, this assay detects p24 from HIV-1 clades A-F | |
2°C to 8°C |
5 x 96 Microplate Wells | |
5 x 96 Determinations |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only. Not for in vitro diagnostic use.