Learn More
ZeptoMetrix RETROtek Horse IgG ELISA
Measures horse IgG in horse serum, plasma or other biological fluids
Supplier: ZeptoMetrix 0801221
Includes: One 96 well microplate, 12mL detector antibody, 7mL of Horse IgG standard, 100mL assay diluent, 125mL of plate wash buffer, 12mL substrate, 12mL stop solution, 1 pack of microplate sealers , 1 plastic bag.
Description
- Contains ready–to–use reagents
- Takes less than two hours to perform
- Microplate and detector antibody reacts uniformly with all subclasses of horse IgG
Specifications
RETROtek Horse IgG ELISA kit | |
Microplate wells coated with polyclonal antibodies to horse IgG form the capture phase of the assay, these antibodies bind uniformly to all subclasses of horse IgG, captured horse IgG then reacts with detector antibody, which is a polyclonal anti-horse IgG, conjugated with horseradish peroxidase | |
2°C to 8°C |
96-well Microplate | |
96 Determinations |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only. Not for in vitro diagnostic use.