Learn More
ZeptoMetrix Corporation SeroDetect QC Validation Panels
For monitoring assay performance across wide range of reactivity levels
Manufacturer: ZeptoMetrix Corporation KZMC012
Alert:
Product capable of transmitting infectious agents.
Description
- Prepared from proteins derived from human sources, and non human components (antimicrobial agents and stabilizers)
- Processed to eliminate contaminants
- Needs evaluation of members as per instructions
CMV IgG Range Validation Panel
- Three vials SeroDetect CMV IgG Negative (1.5mL each)
- Seven vials SeroDetect CMV IgG Positive (1.5mL each)
- One vial SeroDetect CMV IgG Negative (1.5mL each)
- Four vials SeroDetect CMV IgG Positive (1.5mL each)
- One vial SeroDetect HIV positive material (1.25mL)
- One vial SeroDetect HBsAg positive material (1.25mL)
- One vial SeroDetect HBc Total positive material (1.25mL)
- One vial SeroDetect HBc IgM positive material (1.25mL)
- One vial SeroDetect Negative material (1.25mL)
- One vial SeroDetect HIV, non-reactive for antibodies and antigens to HIV (1.25mL)
- Four vials SeroDetect HIV, containing antibodies and antigens to HIV (1.25 each)
Not substitute for mandatory positive and negative control reagents provided with licensed test kits.
For training, lot-to-lot comparison of reagent test kits and to evaluate and compare intra laboratory and inter laboratory performances
Specifications
2 | |
2° to 8°C | |
In vitro assay procedures for the determination of Antibodies to Hepatitis C Antigen (Anti-HCV) | |
5 x 1.25mL | |
Vial |
SeroDetect HIV Ag/Ab Combo Panel | |
SeroDetect HIV Ag/Ab combo panel | |
60 Days | |
Solution |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.