ZeptoMetrix Corporation SeroDetect™ QC Validation Panels Available on GSA/VA Contract for Federal Government customers only.

For monitoring assay performance across wide range of reactivity levels

Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner KZMC013

 View more versions of this product

Catalog No. 22-157-300

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Alert

Product capable of transmitting infectious agents.


    Specifications

    Biosafety Level 2
    For Use With (Application) In vitro assay procedures for the determination of Antibodies to Hepatitis C Antigen (Anti-HCV)
    Format Solution
    Media Type Vial
    Product Type SeroDetect mixed titer panel
    Quantity 5 x 1.25mL
    Shelf Life 60 Days
    Storage Requirements 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit