Learn More
ZeptoMetrix Corporation Viral Lysate, Influenza A H1N1 2009 (Swine Flu)
Can be utilized as an antigen, as a source for the purification of viral proteins, or for the detection of viral antibodies
Manufacturer: ZeptoMetrix Corporation 0810109J
Description
- Propagated in the MDCK cell line
- Purified using sucrose density gradient ultracentrifugation, disrupted in the presence of 0.5% Triton X-100 non-ionic detergent/0.6M KCl, and heat inactivated
Specifications
2 | |
Canada isolate | |
Influenza A 2009 H1N1 (Swine Flu) virus Canada/6294/09 strain purified viral lysate | |
1 Year | |
Solution | |
ISO, 13485 |
Viral Antibodies | |
-65°C | |
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay | |
1mg | |
Vial |