Learn More
ZeptoMetrix Corporation Viral Lysate, Varicella-Zoster Virus
Non-infectious viral lysate purified by sucrose gradient centrifugation prior to inactivation
Manufacturer: ZeptoMetrix Corporation 0810175
Description
This product does contain material derived from human sources and should be handled cautiously as a hazardous material according to good laboratory practices.
Specifications
Viral Antibodies | |
-65°C | |
Dot blotting, Immunodetection of Antibodies to Adenovirus using solid phase Enzyme immunoassays, western blotting, Other Protein based assay | |
Solution | |
ISO, 13485 |
Isolate D | |
Varicella Zoster virus isolate D purified viral lysate | |
1mg | |
Vial |