Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Corporation West Nile Virus Culture Fluid

Virus preparation containing live whole organisms for various research and product development needs

Specifications

Certifications/Compliance ISO, 13485
Format Solution
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Media Type Vial
Quantity 1mL
View More Specs

Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
22-157-583
View Documents
ZeptoMetrix Corporation
0810033CF
West Nile virus NYS-1 (6263) strain infectious culture fluid
Each for $306.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-637
View Documents
ZeptoMetrix Corporation
0810081CF
West Nile virus 956-99 strain infectious culture fluid
Each for $306.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-639
View Documents
ZeptoMetrix Corporation
0810082CF
West Nile virus 2 (1986) strain infectious culture fluid
Each for $306.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

IMPORTANT: Contains live material, so cust must submit an executed ZeptoMetrix MTA before vendor can ship. Also, a USDA APHIS VS 16-6 or 16-6A authorized permit must be obtained and a valid copy of the permit received by ZeptoMetrix prior to shipment of this product. requests.

Specifications

Specifications

ISO, 13485
Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
1mL
-65°C
Solution
Vial
1 Year
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit