Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix ZeptoBind Immunoassay Stabilization Reagent

Ensures binding of protein matrices to the plates

Specifications

Type Immunoassay Stabilization Reagent
Intended Use To aid in the adherence of Antigens or Antibodies to microwell plates
Storage Requirements 4°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
22-157-362
View Documents
ZeptoMetrix
0801184
60mL
Each for $51.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
22-157-490
View Documents
ZeptoMetrix
0801184L
1000mL
Each for $226.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Aids in the adherence of antigens or antibodies to microwell plates
  • Liquid, ready–to–use format
  • Contains no preservatives

Use aseptic technique when removing contents.

Specifications

Specifications

Immunoassay Stabilization Reagent
To aid in the adherence of Antigens or Antibodies to microwell plates
4°C
SDS

Research purposes only. Not for in vitro diagnostic use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit