ZeptoMetrix Corporation ZeptoCoat Immunoassay Stabilization Reagent Available on GSA/VA Contract for Federal Government customers only.
Designed to protect and extend the shelf life of immobilized antigen and antibody matrices applied to microwell plates
Manufacturer: ZeptoMetrix CorporationSupplier Diversity Partner 0801179
 View more versions of this product
Catalog No. 22-157-360
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
Disclaimers:
Research purposes only. Not for in vitro diagnostic use.
    Specifications
    Product Type Polymeric immunoassay stabilization reagent
    For Use With (Application) To protect and extend the shelf life of immobilized Antigen and Antibody matrices applied to microwell plates, increases product shelf life through protection of the Protein matrices
    Storage Requirements 4°C
    Quantity 60mL,60mL
  • Description & Specifications
Related Products
Product Content Feedback