Learn More
Meccanica GM Divisione Medicale ZERO ZERO Prefilled Formalin Container, 60 mL
Meccanica GM Divisione Medicale ZERO Prefilled Formalin Container, 60 mL protects against exposure to harmful chemicals and vapors during routine fixation of specimens in formalin.
$411.93
Specifications
Volume (Metric) 60 mL
View More Specs
Products 8
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-906-016
View Documents
Meccanica GM Divisione Medicale ZERO
6016
Green
Case of 192 for $411.93
Add to cart
22-906-015
View Documents
Meccanica GM Divisione Medicale ZERO
6015
Pink
Case of 192 for $411.93
Add to cart
22-906-013
View Documents
Meccanica GM Divisione Medicale ZERO
6013
Blue
Case of 192 for $411.93
Add to cart
22-906-011
View Documents
Meccanica GM Divisione Medicale ZERO
6011
TRL
Case of 192 for $411.93
Add to cart
22-906-012
View Documents
Meccanica GM Divisione Medicale ZERO
6012
Orange
Case of 192 for $411.93
Add to cart
22-906-014
View Documents
Meccanica GM Divisione Medicale ZERO
6014
Yellow
Case of 192 for $411.93
Add to cart
22-906-017
View Documents
Meccanica GM Divisione Medicale ZERO
6017
Grey
Case of 192 for $411.93
Add to cart
22-906-018
View Documents
Meccanica GM Divisione Medicale ZERO
6018
Lilac
Case of 192 for $411.93
Add to cart
Description
Description
  • Reduces or eliminates exposure to formalin and harmful vapors
  • Comes in a variety of colored caps for easy specimen identification
  • Security seal
  • Dual chamber keeps formalin trapped until container is activated
Specifications
Specifications
60 mL