Learn More
Zero Solution 0.4% Hydroxylamine Hydrochloride Low Mercury, Reagents
Specifications
Specifications
| Chemical Name or Material | Zero Solution 0.4% Hydroxylamine Hydrochloride Low Mercury |
| Packaging | Cubitainer |
| CAS | 7732-18-5 |
| Specific Gravity | 1.0034 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solubility | Completely miscible |
| Quantity | 10 L |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.