Learn More
Medchemexpress LLC Zeteletinib | 2216753-97-6 | 98.3% | C25H23F3N4O4 | 10MG

Supplier: Medchemexpress LLC HY13959010MG
Zeteletinib is an orally active, selective RET kinase inhibitor supplied as a white-to-light-yellow solid for research use. The compound shows nanomolar potency against RET and retains activity against clinically relevant RET mutants; documentation includes solubility data and recommended storage for stability.
- Orally active, selective RET kinase inhibitor.
- Nanomolar potency against RET and activity against RETV804M/L and M918T mutants.
- High purity (98.3%) suitable for research applications.
- Solid form, white to light yellow.
- Soluble in DMSO (100 mg/mL); ultrasonic recommended.
- Recommended storage: powder at -20°C (long term) or 4°C; in solvent at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.