Learn More
Abnova™ ZFAND1 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00079752T03
Catalog No. 89-967-814
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZFAND1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 18.2kDa
Transient overexpression cell lysate was tested with anti-ZFAND1 antibody (Mfr. No. H00079752-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC056877
pCMV-ZFAND1 full-length
100 μL
Human