Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND2B Mouse anti-Human, Clone: OTI2G10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502514

Catalog No. 50-167-3567


Add to cart

Description

Description

This gene encodes a protein containing AN1-type zinc-fingers and ubiquitin-interacting motifs. The encoded protein likely associates with the proteosome to stimulate the degradation of toxic or misfolded proteins. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

ZFAND2B
Monoclonal
0.47 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WV99
AIRAPL
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
130617
Flow Cytometry, Western Blot
OTI2G10
Unconjugated
ZFAND2B
Liquid
ZFAND2B
Full length human recombit protein of human ZFAND2B produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit