Learn More
Aviva Systems Biology Corp ZFAND3 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP34469_P050
This is a rabbit polyclonal antibody against ZFAND3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZFAND3. Alternate names: FLJ13222, TEX27ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55407