Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND3 Mouse anti-Human, Clone: OTI1C5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526352

Catalog No. PIMA526352


Add to cart

Description

Description

ZFAND3 (Zinc Finger AN1-Type Containing 3) is a Protein Coding gene and also known as Testis expressed sequence 27.
Specifications

Specifications

ZFAND3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H8U3
AN1-type zinc finger protein 3; Testis-expressed sequence 27; ZFAND3; TEX27
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
60685
Immunohistochemistry (Paraffin), Western Blot
OTI1C5
Unconjugated
ZFAND3
Liquid
ZFAND3
Full length human recombit protein of ZFAND3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit