Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFAND5 Mouse anti-Human, Clone: OTI1D8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526455

Catalog No. PIMA526455


Add to cart

Description

Description

ZNF216 (ZFAND5) is a potent inhibitory factor for osteoclast differentiation and the mechanism is unlikely due to direct attenuation of the NF-kappa B pathway. It also has redundant and distinct roles in regulating apoptosis.
Specifications

Specifications

ZFAND5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O76080
ZFAND5, ZA20D2, ZFAND5A, ZNF216
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7763
Immunohistochemistry (Paraffin), Western Blot
OTI1D8
Unconjugated
ZFAND5
Liquid
ZFAND5
Full length human recombit protein of ZFAND5 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit