Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZFAND6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540895
Description
Peptide sequence: SDTAQQPSEE QSKSLEKPKQ KKNRCFMCRK KVGLTGFECR CGNVYCGVHR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
ZA20D3 is a zinc-finger protein located on chromosome 15.
Specifications
ZFAND6 | |
Polyclonal | |
Unconjugated | |
ZFAND6 | |
3110005P07Rik; AN1-type zinc finger protein 6; associated with PRK1 protein; associated with Prkcl1; Awp1; HT032; protein associated with PRK1; Za20d3; ZFAND5B; ZFAND6; zgc:101121; zinc finger A20 domain-containing protein 3; zinc finger AN1-type containing 6; zinc finger, A20 domain containing 3; zinc finger, AN1-type domain 6 | |
Rabbit | |
Protein A | |
RUO | |
54469 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6FIF0 | |
ZFAND6 | |
Synthetic peptide directed towards the middle region of human ZA20D3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction