Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP106 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP288616

Catalog No. NB036238


Add to cart

Description

Description

ZFP106 Polyclonal specifically detects ZFP106 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZFP106
Unconjugated
PBS, 2% Sucrose
DKFZp451A239, FLJ34610, SH3BP3, SH3-domain binding protein 3, Zfp-106, zinc finger protein 106 homolog, zinc finger protein 106 homolog (mouse), Zinc finger protein 474, ZNF474FLJ45841
The immunogen is a synthetic peptide directed towards the N-terminal region of human ZFP106
100 μg
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Immunogen affinity purified
RUO
Primary
64397
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit