Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Zfp157 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537973
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Specifications
Zfp157 | |
Polyclonal | |
Unconjugated | |
Zfp157 | |
2610020C11Rik; A630094N24Rik; AI327407; mszf12; mszf23-1; Zfp157; zinc finger protein 157 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
72154 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
0 | |
Zfp157 | |
Peptide with sequence QCSKQHAYYPSVKP, from the internal region of mouse Zfp157. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction